Learn More
Stoko™ Stoko™ Kresto™ Classic Cleanser
Quickly and effectively cleans grease, oil, grime, ink, carbon black, sewer sludge, adhesives, and other industrial dirt
Manufacturer:  Stoko™ 9 87045 06
 View more versions of this product
Catalog No. 19-819-820
Add to cart
Description
Description
  • Concentrated formula with ASTOPON™ natural walnut shell powder scrubber
  • Eucornol™ for extra skin protection
  • Gentle low-solvent formulation
  • Pleasant fragrance and pH balanced
  • Completely biodegradable
  • Will not clog drains or pipes
Specifications
Specifications
68 oz.
Bottle, soft
Water, C13-14 Isoparaffin, Walnut Shell Powder, Sodium Laureth Sulfate, Sulfated Castor Oil, Sodium Chloride, Cocamidopropyl Betaine, Quaternium 18 Bentonite, Cellulose Gum, Titanium Dioxide, Citric Acid, Fragrance, Potassium Sorbate, 2-Bromo-2-Nitropropane-1,3-Diol
Kresto™ Cleanser
Pleasant
5.2 to 5.8
2L
6 Case
Light beige
Paste
Quickly and effectively cleans grease, oil, grime, ink, carbon black, sewer sludge, adhesives, and other industrial dirt