Stoko™ Kwik-Wipes™ Towelettes

Perfect when a sink is unavailable or at individual work stations

Manufacturer: Stockhausen Inc. 2 87077 06

 View more versions of this product

Catalog No. 19-141-623

  Check Availability
Add to Cart

    Specifications

    Color White
    Product Type Kwik-Wipes Towelette
    Packaging Bucket
    Quantity 4 Case
    Composition Water, Dipropylene Glycol Methyl Ether, Sodium Caprylyl Sulfonate, Nonoxynol-10, Sodium Laureth Sulfate, PEG-75 Lanolin, fragrance, 2-Bromo-2-Nitropropane-1,3-Diol
    For Use With (Application) For use on skin, also great for tools and other surfaces, quick cleaning of dirt, grease, and grime from hands
    Format Wipe
    Shelf Life 12 Month(s)
    pH Range 6

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit