Learn More
Stoko™ Skin Care Arretil™ Before Work Cream
Manufacturer:  Stoko™ 33865
Catalog No. 19-824-002
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
Description
Description
Specifications
Specifications
Before Work Cream
Simplifies the removal of epoxy and resins from the skin; Ideal when working with grease, oil, cutting oils, and organic solvents
Tube
Lotion
100 mL