Learn More
Stoko™ Skin Care Reduran™ Special Cleaner
Manufacturer:  Stockhausen Inc. KKU250ML
 View more versions of this product
Catalog No. 19-824-004
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
Removal of dyestuffs
Contains a reducing agent and synthetic detergents that emulsify the dye in water; Contains all-natural walnut shell scrubbers
Skin Cleanser
100mL tube