Stoko™ Skin Care Reduran™ Special Cleaner
Manufacturer: Stockhausen Inc. 33885
 View more versions of this product
Catalog No. 19-824-005
  Check Availability
Add to Cart
    Specifications
    Size (Metric) 1000mL softbottle
    For Use With (Application) Removal of dyestuffs
    Format Contains a reducing agent and synthetic detergents that emulsify the dye in water; Contains all-natural walnut shell scrubbers
    Product Type Skin Cleanser
  • Description & Specifications
Related Products
Product Content Feedback