Learn More
Stoko™ Skin Care Travabon™ Before Work Cream
Cream helps protect skin from oil-based hazards
Manufacturer:  Stoko™ 33866
 View more versions of this product
Catalog No. 19-824-018
Add to cart
Description
Description
For organic solvents, paint thinner, oil, paint, ink, and metal dust.
  • Contains special, skin-compatible emulsifiers which assist in cleanup of heavy, stubborn dirt
  • Reduces the adherence of non-water-soluble substances like grease, oil, glues, resins, paints and graphite to the skin
  • Forms a protective layer that works to reduce the penetration of non-water-soluble contaminants through the top layers of the skin
  • Contains no silicone that can cause problems in sensitive coating operations
Order Info
Tube size: 3.4 oz. (100mL). Also available in 34 oz. (1000mL) Softbottle. Contact your Fisher Safety Customer Service Representative for ordering information.
Specifications
Specifications
3.3 oz., 100mL
Befor work cream
100mL tube