Learn More
Stoko™ Solopol™ Classic Heavy-Duty Hand Cleanser
Solvent-free cleanser quickly and effectively cleans grease, grime, oil, and other medium to heavy industrial dirt
Manufacturer:  Stockhausen Inc. 9 83187 06
 View more versions of this product
Catalog No. 19-141-629
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
Light beige
Cleans grease, grime, oil, and other medium to heavy industrial dirt
Solopol™ Classic Hand Cleanser
68 oz.
Bottle, soft
Water, Walnut Shell Powder, Sodium Laureth Sulfate, Sulfated Castor Oil, Cocamidopropyl Betaine, Sodium Chloride, Oleic Acid, Citric Acid, Titanium Dioxide, Cellulose Gum, Xanthan Gum, Fragrance, Citric Acid, Potassium Sorbate, 2-Bromo-2-Nitropropane-1,3-Diol
Paste
4.9 to 9.3
2L
6 Case
Safety and Handling
ShelfLife : 30 Month(s)