Learn More
Stoko Spray Hand Sanitizer
Manufacturer:  Stockhausen Inc. 5 50100 12
 View more versions of this product
Catalog No. 19-130-1993
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
Stoko Spray™ Moisturizing Cleanser
Light Green
Liquid
400mL
5.3
Water, Sodium Laureth Sulfate, Sodium Cocoamphoacetate, Glycerin, Sodium Chloride, Citric Acid, PEG-6 Caprylic/Capric Glycerides, Sodium Benzoate, Glycol Distearate, Fragrance, Steareth-4, Dye
13.5 oz.
12 Case