Stone Medical Clean Collect™ Blood Culture Kits

Blood culture diversion system prevents contamination in peripheral blood cultures

Manufacturer: STONE MEDICAL CORPORATION CC225B

 View more versions of this product

Catalog No. 23-200-105

    N/A

Due to product restrictions, we cannot sell this product online. Please call Customer Service at 1-800-766-7000 or send an email to CS.WebEmail@thermofisher.com for assistance. 


    Specifications

    Item Description Diversion tube, blood culture bottle adaptor, Prevantics™ Swabstick antiseptic, sterile gloves, tourniquet, alcohol pads, bandage, instruction card, tracking decals
    Description Blood Culture Kit
    Storage Requirements Ambient temperature (0-40°C) dry storage
    Includes Diversion tube, blood culture bottle adaptor, Prevantics Swabstick antiseptic, sterile gloves, tourniquet, alcohol pads, bandage, instruction card, tracking decals
    For Use With (Application) Compatible with BacT/Alert Systems

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit