Learn More
Clean Collect™ Blood Culture Kits
Blood culture diversion system prevents contamination in peripheral blood cultures
Supplier:  Stone Medical Corporation CC220R
 View more versions of this product
Catalog No. 23-200-102
Item Discontinued Due to product restrictions, please to purchase or view availability for this product.This item has been discontinued by the supplier and is no longer available. Please reach out to your sales representative.
Description
Description
Reducing blood culture contamination rates prevents unnecessary treatment of patients. Clean Collect kits can help to increase quality and save considerable resources.
  • Prevents blood culture contamination using proprietary diversion method
  • Average contamination rate of 0.5% compared to 3% national average
  • Diversion method reduces contamination rates by an average of 74%
  • Standardized kits promote consistency for long term improvement
  • Reduces hospital costs considerably
  • Ambient temperature (0-40°C) dry storage; 10 month shelf life
Specifications
Specifications
Ambient temperature (0-40°C) dry storage
Diversion tube, BD push button butterfly, standard needle hub, Prevantics™ Swabstick antiseptic, sterile gloves, tourniquet, alcohol pads, bandage, instruction card, tracking decals