Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Clean Collect™ IV Blood Culture Kits

Prevents blood culture contamination using proprietary diversion method. Standardized kits promote consistency for long-term improvement.
Catalog Number | Mfr. No. | Description | Includes | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Description | Includes | Price | Quantity & Availability | ||||
23-200-103
![]() |
LSL Healthcare Inc
CC310 |
Compatible with Bactec or Versatrek Systems | Diversion tube, IV blood collection device, Chloraprep™ Frepp™ antiseptic, sterile gloves, tourniquet, alcohol pads, Suresite IV dressing, Curad tape roll, gauze, instruction card, tracking decals |
Case of 100 for $1,189.32
|
N/A
![]() |
||||
Due to product restrictions, please Sign In to purchase or view availability for this product. | |||||||||
23-200-107
![]() |
LSL Healthcare Inc
CC315 |
Compatible with BacT/Alert Systems | Diversion tube, IV luer adaptor, blood culture bottle adaptor, Chloraprep™ Frepp™ antiseptic, sterile gloves, tourniquet, alcohol pads, Suresite IV dressing, Curad tape roll, gauze, instruction card, tracking decals |
Case of 40 for $369.24
|
N/A | ||||
Due to product restrictions, please Sign In to purchase or view availability for this product. | |||||||||
Description
- Blood culture diversion system prevents contamination in IV drawn blood cultures
- Average contamination rate of 0.5% compared to 3% national average
- Diversion method reduces contamination rates by an average of 74%
- Reduces hospital costs considerably
- Ambient temperature (0 to 40°C) dry storage; 10 month shelf life
Specifications
Ambient temperature (0-40°C) dry storage |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title