Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Synthware™ Cap Style Stopper
For an apparatus with an inner opening.
$21.00 - $29.00
Specifications
Color | Clear |
---|---|
Material | Borosilicate Glass |
For Use With (Application) | Organic Synthesis |
Quantity | 1 Ea. |
Catalog Number | Mfr. No. | Size | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Size | Price | Quantity & Availability | |||||
31-501-889
|
Synthware™
S391420 |
14/20 |
Each for $21.00
|
|
|||||
31-501-890
|
Synthware™
S392440 |
24/40 |
Each for $23.75
|
|
|||||
31-501-891
|
Synthware™
S392942 |
29/42 |
Each for $29.00
|
|
|||||
Specifications
Clear | |
Borosilicate Glass | |
Organic Synthesis | |
1 Ea. |
Spot an opportunity for improvement?
Provide Content Correction
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title