Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

DWK Life Sciences Wheaton™ Stoppers for Serum Bottles and Vials: Straight Plug Stopper

Provide airtight covers

Manufacturer:  DWK Life Sciences 224100330

 View more versions of this product

Catalog No. 06-451-61


Add to cart

Description

Description

  • Flange-type stoppers are secured with aluminum crimp seals
  • Slotted and three-prong plug stoppers are for freeze-dry applications
Specifications

Specifications

3cm
40 Durometer
Red
Rubber
Fits 1.5cm I.D., 3cm O.D. mouth of serum bottles and vials
Yes
30mm
SDS
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit