Learn More
Saf T Pak Inc STP-270 - Category B Ambient Shipping System, (UN3373)
Supplier: Saf T Pak Inc STP270
STP-270 - Category B Ambient Shipping System, (UN3373), Outer Box – OD 15.5 x 12.5 x 5in., pre-printed with: Biological Substance, Category B, UN 3373 Marking, Exempt Human/Animal Specimen Marking, Orientation Arrows, Designed for shipments of large specimens or multiple specimens in a single package, 25/Case