Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

STRA8 Rabbit anti-Human, Polyclonal, Invitrogen

Rabbit Polyclonal Antibody

Manufacturer:  Invitrogen PA551126

Catalog No. PA551126


Add to cart

Description

Description

STRA8 Polyclonal Antibody detects endogenous levels of STRA8 protein.

Meiosis-inducer required for the transition into meiosis for both female and male germ cells. In female germ cells, required for premeiotic DNA replication and subsequent events in meiotic prophase. During spermatogenesis, next to its role in meiotic initiation, promotes (but is not required for) spermatogonial differentiation. Can associate with DNA (possibly in an indirect manner), and in vitro can activate DNA transcription. {ECO:0000269; PubMed:16461896, ECO:0000269; PubMed:17115059, ECO:0000269; PubMed:18799751, ECO:0000269; PubMed:18799790, ECO:0000269; PubMed:19805549, ECO:0000269; PubMed:25902548}.
Specifications

Specifications

STRA8
Polyclonal
Unconjugated
STRA8
Liquid
STRA8
Synthesized peptide derived from the Internal region of human Stra8.
Antigen affinity chromatography
RUO
Antibody
Polyclonal
Human, Mouse
ELISA, Western Blot
1 mg/mL
PBS with 0.5% BSA, 50% glycerol and 0.02% sodium azide; pH 7.4
Q7Z7C7
STRA8, Stimulated by retinoic acid gene 8 protein homolog
Rabbit
IgG
100 μL
-20° C, Avoid Freeze/Thaw Cycles
Primary
346673
Documents
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit