Learn More
Cole-Parmer Masterflex Fitting, PVDF, Straight, Hose Barb Union, 1/4" ID; 10/PK  DFS Item
Manufacturer:  Cole-Parmer 3061216
These economical straight barbed fittings easily connect fluid paths without the use of tools. Just slip your flexible tubing or hose onto the barb to create a secure seal.
Catalog No. 50-629-218