Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
DWK Life Sciences Wheaton™ Straight Sided Glass Bottle
Click to view available options
No. per Case:
12/Case
24/Case
48/Case
Description
- Straight smooth walls allow for complete removal of contents
- Ideal for soil sampling and environmental applications
- With attached screw caps lined in PTFE or PV
Specifications
Closure Liner | PTFE |
Closure Material | Polypropylene |
Product Type | Straight Sided Bottle |
Capacity (Metric) | 125 mL |
Color | Amber |
Material | Borosilicate Glass (Type 3) |
No. per Case | 24/Case |
Closure Color | White |
Mouth | Wide |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction