Learn More
Alfa Aesar™ Straight Wall Inconel Crucible
Manufacturer:  Alfa Aesar™ 36099KT
 View more versions of this product
Catalog No. AA36099KT
Order in Bulk
Request bulk or custom formats
Add to cart
Description
Description
Specifications
Specifications
Straight Wall
Outside: 51mm
51mm x 41mm
75mL
Inconel
Safety and Handling
Safety and Handling
Recommended Storage : Ambient temperatures