Learn More
Alfa Aesar™ Straight Wall Zirconium Crucible
Manufacturer:  Alfa Aesar™ 35992KT
 View more versions of this product
Catalog No. AA35992KT
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
35mL
89mm
Zirconium
Outside: 46mm
Straight Wall
Zirconium Crucible