Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Streamlight™ 3C ProPolymer™ Lux Flashlight

Features a deep parabolic reflector that produces long range brilliance and a smooth uniform light pattern

$93.00

Specifications

Battery C Alkaline
Certifications/Compliance UL Class I, Division 1, Class II, Division 1 and Class III, T4
Controller Type Rubber booted, tail-end push button-switch
Includes White LED
Length (English) 9.3 in.
View More Specs

Products 1
Catalog Number Mfr. No. Description Price Qty Availability  
Catalog Number Mfr. No. Description Price Qty Availability  
19-819-889 View Documents Streamlight™
33702
N/A Color: Black Each for $93.00
Add to cart
 
Description

Description

  • 10,000 Hour, 1 watt super high-flux lux LED
  • Up to 3500 candela, 44 lumens
  • Powered by three “C” alkaline batteries, not included
  • Runs up to four hours at full intensity; up to 15 hrs to 50 percent output intensity
  • Rubber booted, tail-end push button-switch
  • Momentary and constant on
  • Spring-loaded clip
  • Water resistant, O-ring sealed (not dive rated)
  • Corrosion proof and non-conductive
  • Unbreakable polycarbonate lens with scratch resistant coating
  • Shock resistant

Order Info

Length: 9.3 in. (23.6cm). Weight (with batteries): 14.6 oz. (414g).

Certifications

UL listed for Class I, Division 1; Class II, Division 1; and Class III hazardous locations; Exia Temperature Code T4.

Specifications

Specifications

C Alkaline
Rubber booted, tail-end push button-switch
9.3 in.
LED
Polymer
15 oz.
UL Class I, Division 1, Class II, Division 1 and Class III, T4
White LED
23.6cm
44lm
LED Flashlight
0.42kg
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit