Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Streamlight™ 4AA ProPolymer™ Lux Flashlight
The 4AA ProPolymer Lux is a durable polymer flashlight that provides a longer reaching beam and is ergonomically shaped for maximum hand comfort.
Supplier: Streamlight™ 68202
Includes: Wrist lanyard
Description
High performance 4AA flashlight features a body constructed of a polymer engineering resin that is virtually indestructible, shock resistant, non-conductive and lightweight.
- One C4™ LED with a 50,000 hour lifetime
- Deep reflector produces long-range brilliance and smooth beam
- 4200 candela, 100 lumens
- Push-button tail switch for one-handed operation
- Impact-resistant polymer body
- Unbreakable polycarbonate lens with scratch resistant coating
- O-ring sealed, water resistant
- Corrosion proof and non-conductive
- Powered by four “AA” alkaline batteries, included
- Four hours at full intensity, two additional hours of declining usable light
- Power regulated for maximum output throughout battery life
Certifications
UL listed for Class I, Division 1; Class II, Division 1; and Class III hazardous locations
Specifications
Flashlight | |
100,000 hr. | |
0.173 kg | |
155 hr. | |
CE and MSHA, UL Class I, Div. 1, Groups C, D; Class I, Div 2, Groups A, B, C, D; Class II, Div. 2, Groups F&G; Class III, T3C | |
Wrist lanyard connection | |
16.5 cm | |
Polymer |
C4 LED | |
6.3 oz. | |
155 hr. | |
Alkaline | |
Yellow or Black | |
6.5 in. | |
67 lm |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction