Learn More
Streamlight™ 61080 Trident HP Headlamp
Manufacturer:  Streamlight™ 61080
Catalog No. 19-500-559
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.
View substitute products »
Description
Description
  • Unbreakable polycarbonate lens
  • High power LED. LED rating: 2000 candela peak beam intensity; 50-Lumen
  • Water resistant lightweight thermoplastic construction
  • Uses three (3) “AAA” alkaline batteries
Specifications
Specifications
Safety