Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Streamlight™ PolyStinger™ Flashlight
Suitable for use in marine, damp, and corrosive environments
Supplier: Streamlight™ 76014
Includes: Spare bulb in tailcap and AC/DC chargers and holders
Description
- Powerful, rechargeable flashlight with Xenon bi-pin bulb provides up to 11,000 candela with 90 minutes of continuous run time between charges
- Recharges on 110VAC or 12VDC steady chargers with 10-hour charge time using
- Fast charger enables 2.5 hour charge time
- Constructed of yellow, super-tough, nonconductive polymer with rubber nonslip grip
- Compact snap-in charger recharges NiCd battery up to 1000 times; LED light indicates when flashlight is fully charged
Order Info
Length: 7.34 in. (18.7cm). Weight: 8.7 oz. (247g).
Certifications
UL listed for certain Class I, Division 2 hazardous environments; CE marked.
Specifications
Rechargeable Flashlight | |
110 V | |
0.847 kg | |
Yellow | |
Spare bulb in tail cap and AC/DC chargers and holders | |
18.4 cm |
Xenon | |
8.7 oz. | |
UL, CE, Class I, Div. 2, Groups A,B,C,D T1; Class II, Div. 2, Group G T3 | |
For use in marine, damp, and corrosive environments | |
7.34 in. | |
Nylon Polymer |
Provide Content Correction
We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?
Provide Content Correction