Streamlight™ Rechargeable LiteBox™ Lanterns

Adjustable swivel neck allows for rotation of the lamp head

Manufacturer: Streamlight 45102

 View more versions of this product

Catalog No. 18-999-329

  Check Availability
Add to Cart

    Specifications

    Battery Type 12AH
    Certifications/Compliance NFPA 1901-12-1.7
    For Use With (Application) Fire and rescue, industrial safety, utilities, emergency medical services, homeowners, sporting goods, pest control
    Height (English) 7 in.
    Height (Metric) 17.8cm
    Light Source High-intensity halogen bi-pin bulb in 8 or 20 watt spot for 8- or 20 watt bulbs, 3.5 for 20 watt
    Lighting Type LED
    Lumens 400
    Material ABS Thermoplastic
    Style Swivel neck, Vehicle-Mount System
    Wattage 20w
    Weight (Metric) 3.3kg
    Color Orange
    Product Type Safety Lantern
    Includes Vehicle-mount hard-wire rack, shoulder carrying strap
    Length (English) 11.5 in.
    Length (Metric) 29.2cm
    Weight (English) 7.4 lb.

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit