Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Streamlight™ Vulcan™ Rechargeable Lantern Systems
SDP

Powerful lantern comes in fire service version with LED tail lights

$230.00 - $257.50

Specifications

Battery Type Zero-maintenance 4.5 Ah sealed lead acid
Burn Time 3.25 hr.
Certifications/Compliance Class l, ll, Ill, Division 2, NFPA 1901-12-1.7, Class I, II, Division 2, Class III
Material ABS Thermoplastic Housing
Weight (English) 3.3 lb.
View More Specs

Products 3
Catalog Number Mfr. No. Color Includes Lumens Price Quantity & Availability  
Catalog Number Mfr. No. Color Includes Lumens Price Quantity & Availability  
19-041-388
SDP
View Documents
Streamlight™
44200
Yellow Shoulder strap, AC and DC chargers 157 lm
Each for $252.00
Add to cart
 
19-121-807
SDP
View Documents
Streamlight™
44400
Orange Ultrabright taillight LEDs 150 lm
Each for $257.50
Add to cart
 
19-186-170
SDP
View Documents
Streamlight™
44000
Yellow Shoulder strap and charging/mounting rack, AC/DC chargers 157 lm
Each for $230.00
Add to cart
 
Description

Description

System Components

  • Rechargeable flashlight with cushioned grip handle and a high-impact, waterproof, ABS thermoplastic housing
  • 6V rechargeable battery
  • 120VAC and 12VDC power cords
  • Halogen bi-pin bulb
  • 80,000 candela
  • Charging/mounting rack, which meets requirements of NFPA 1901-12-1.7 (1999) mounted in any position
  • Shoulder strap
Fire Vulcan™ Option
  • Extremely powerful (65,000 candlepower) fire service flashlight features two ultrabright blue tail light LEDs that allow you to see others in smoke-filled rooms
  • With quick-release strap and glove-friendly, three-position ON/OFF toggle switch

Order Info

Maximum candlepower with supplied 8w spot: up to 80,000. Run time: 3.25 hr. on regular Vulcan; Fire Vulcan option provides 3 hours. Weight: 3.3 lb. (1.5kg). Length: 7.5 in. (19cm).

Certifications

USTC approved for Class l, ll, lll, Division 2. Most models meet applicable European Community Directives, including ATEX Zone 2

Compliance

Rack meets requirements of NFPA 1901-12-1.7 mounted in any position.

Warranty and Services

Limited lifetime

Specifications

Specifications

Zero-maintenance 4.5 Ah sealed lead acid
Class l, ll, Ill, Division 2, NFPA 1901-12-1.7, Class I, II, Division 2, Class III
3.3 lb.
3.25 hr.
ABS Thermoplastic Housing
1.5 kg
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit