Streck CD-Chex™ CD34

Positive procedural control for enumerating CD34 positive cells by flow cytometry

Manufacturer: Streck 213382

 View more versions of this product

Catalog No. 23-046-624

  Check Availability
Add to Cart

    Specifications

    Product Type CD34 Flow Cytometry Control
    Stability 30 Days Open, 90 Closed Vial
    For Use With (Application) Used as a complete process control when evaluating CD34 positive cells
    For Use With (Equipment) Flow cytometer
    Shelf Life 3 Months
    Quantity 1 x 1mL
    Level 3

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit