Learn More
Streck CD-Chex™ CD34
Positive procedural control for enumerating CD34 positive cells by flow cytometry
Manufacturer:  Streck 213346
 View more versions of this product
Catalog No. 23-285-519
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
CD34 Flow Cytometry Control
30 Days Open, 90 Closed Vial
Flow cytometer
2
Used as a complete process control when evaluating CD34 positive cells
1 x 1mL
Safety and Handling
ShelfLife : 3 Months