Learn More
Streck CD-Chex™ Plus BC Controls
For immunophenotyping with Beckman Coulter™ flow cytometry systems
Manufacturer:  Streck 213377
 View more versions of this product
Catalog No. 23-046-566
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
CD-Chex™ plus BC control
CD4 Low
3 Months
Beckman Coulter™ flow cytometry system
2° to 10°C
Normal human peripheral blood leukocytes and erythrocytes
Plastic Vial
Determines the accuracy and reproducibility of all steps involved in immunophenotyping
CE
5 x 3mL