Streck CD-Chex™ Plus BC Controls

For immunophenotyping with Beckman Coulter™ flow cytometry systems

Manufacturer: Streck 213378

 View more versions of this product

Catalog No. 23-046-567

  Check Availability
Add to Cart

    Specifications

    Product Type CD-Chex™ plus BC control
    Format Plastic Vial
    Stability 3 Months
    For Use With (Application) Determines the accuracy and reproducibility of all steps involved in immunophenotyping
    For Use With (Equipment) Beckman Coulter™ flow cytometry system
    Certifications/Compliance CE
    Storage Requirements 2° to 10°C
    Quantity 5 x 3mL,5 x 3mL
    Source Normal human peripheral blood leukocytes and erythrocytes,Normal human peripheral blood leukocytes and erythrocytes
    Level Normal,Normal

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit