Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

Streck CD-Chex™ Plus BC Controls
SDP

For immunophenotyping with Beckman Coulter™ flow cytometry systems

Specifications

Certifications/Compliance CE
Type CD-Chex™ plus BC Control
Intended Use Determines the accuracy and reproducibility of all steps involved in immunophenotyping
For Use With (Equipment) Beckman Coulter™ flow cytometry system
Source Normal Human Peripheral Blood Leukocytes and Erythrocytes
View More Specs

Products 7
Catalog Number Mfr. No. Level Quantity Price Quantity & Availability  
Catalog Number Mfr. No. Level Quantity Price Quantity & Availability  
23-046-564
SDP
View Documents
Streck
213375
CD4 Low, Normal 4 x 3 mL This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. View substitute products »
23-046-560
SDP
View Documents
Streck
213371
CD4 Low 1 x 3 mL This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
23-046-565
SDP
View Documents
Streck
213376
CD4 Low, Normal 10 x 3 mL This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. View substitute products »
23-046-562
SDP
View Documents
Streck
213373
Normal 1 x 3 mL This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. N/A
23-046-561
SDP
View Documents
Streck
213372
CD4 Low 2 x 3 mL This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. View substitute products »
23-046-563
SDP
View Documents
Streck
213374
Normal 2 x 3 mL This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. View substitute products »
23-046-567
SDP
View Documents
Streck
213378
Normal 5 x 3 mL This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. View substitute products »
Description

Description

  • Determines the accuracy and reproducibility of all steps involved in immunophenotyping, including red blood cell lysis
  • CD-Chex Plus BC CD4 Low offers a depressed CD4 absolute number often indicated in patients with immunodeficiency diseases
  • Both CD-Chex Plus BC and CD-Chex Plus BC CD4 Low offer 30-day open-vial stability and up to 90-day closed-vial stability
  • Available in plastic vials with pierceable caps containing a 3.0mL fill

Specifications

Specifications

CE
Determines the accuracy and reproducibility of all steps involved in immunophenotyping
Normal Human Peripheral Blood Leukocytes and Erythrocytes
2°C to 10°C
CD-Chex™ plus BC Control
Beckman Coulter™ flow cytometry system
3 Months
SDS
Product Certifications
Provide Content Correction

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit