Learn More
Streck CD-Chex™ Plus BC Controls
For immunophenotyping with Beckman Coulter™ flow cytometry systems
Manufacturer:  Streck 213375
 View more versions of this product
Catalog No. 23-046-564
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
CD-Chex™ plus BC control
CD4 Low, Normal
Determines the accuracy and reproducibility of all steps involved in immunophenotyping
CE
4 x 3mL
Normal human peripheral blood leukocytes and erythrocytes
3 Months
Beckman Coulter™ flow cytometry system
2° to 10°C