Streck CD-Chex™ Plus Immunophenotyping Control

Assayed control for monitoring immunophenotyping by flow cytometry

Manufacturer: Streck 213340

 View more versions of this product

Catalog No. 23-046-502A

  Check Availability
Add to Cart

    Specifications

    Product Type CD-Chex™ plus Immunophenotyping control
    Format Glass Vial
    Stability 3 Months
    For Use With (Application) Immunophenotyping including RBC lysis, Antibody staining, instrument performance and operator technique
    For Use With (Equipment) Beckman Coulter™ flow cytometry system
    Storage Requirements 2° to 10°C
    Quantity 2 x 2.5mL,2 x 2.5mL
    Source Human Peripheral Blood Leukocytes and Erythrocytes,Human Peripheral Blood Leukocytes and Erythrocytes
    Level CD4 Low, Normal,CD4 Low, Normal
    Certifications/Compliance CE

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit