Learn More
Streck CD-Chex™ Plus Immunophenotyping Control Available on GSA/VA Contract for Federal Government customers only.
Assayed control for monitoring immunophenotyping by flow cytometry
Manufacturer:  Streck 213344
 View more versions of this product
Catalog No. 23-046-502C
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
CD-Chex™ plus Immunophenotyping control
CD4 Low, Normal
3 Months
Beckman Coulter™ flow cytometry system
2° to 10°C
Human Peripheral Blood Leukocytes and Erythrocytes
Glass Vial
Immunophenotyping including RBC lysis, Antibody staining, instrument performance and operator technique
CE
10 x 2.5mL