Streck Cell-Chex™ Auto Body Fluid Cell Count Control Available on GSA/VA Contract for Federal Government customers only.

Eliminate the need for manual hemacytometer testing

Manufacturer: Streck 200069

 View more versions of this product

Catalog No. 23-046-623

  Check Availability
Add to Cart

    Specifications

    Product Type Auto body fluid cell count control
    Stability 75 Days Closed, 30 Days Open Vial
    Cap Type Pierceable
    For Use With (Application) Three-level spinal and body fluid control for evaluating the accuracy and precision of hematology instruments that measure Blood cell counts in patient body fluid samples
    For Use With (Equipment) Abbott, Beckman Coulter™, Sysmex™ hematology instruments
    Shelf Life 75 Days
    Storage Requirements 2° to 10°C
    Packaging Vial
    Quantity 3 x 3mL
    Level 3

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit