Streck Cell Preservative™

Manufacturer: Streck 213352

 View more versions of this product

Catalog No. 23-046-501E

  Check Availability
Add to Cart

    Specifications

    Product Type Cell preservative
    pH ≈7.2
    For Use With (Application) Easy to use liquid preservative that stabilizes cell morphology and preserves cellular Antigen expression of WBC for up to 7 days
    For Use With (Equipment) Flow cytometer
    Shelf Life 7 Days
    Storage Requirements 18° to 30°C
    Quantity 24 x 1mL
    Color Clear

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit