Learn More
Streck CVA™ Calibration Verification Assessment/Linearity Kit
Ensuring confidence in every result, CVA (Calibration Verification Assessment) tests the extreme upper and lower limits of an instrument to assure abnormal patient results are valid
Manufacturer:  Streck 291719
 View more versions of this product
Catalog No. 11-716-367
Add to cart
This item is not returnable. View return policy
Description
Description
Assayed linearity control kits to measure the linear performance of of three- and five-part hematology analyzers
  • Instruments include:
  • Beckman Coulter™, Ac-T™ 5diff, HmX, UniCel™ DxH TM800, LH 500, LH 750/755, LH 780/785; HemoCue™, Inc. Hb 201+ Analyzer; Mindray™ BC-3200; Abbott CELL-DYN™ Sapphire™, Ruby™, Emerald™, 3500/3700, 3200, 1800
  • Include customized ranges appropriate for each instrument model and provide instrument-specific assay values for the WBC, platelet, RBC and hemoglobin parameters
  • Open-vial stability is 5 days
  • Closed-vial stability is 120 days
  • All kits include WBC, platelet and RBC/hemoglobin vials unless otherwise noted
Specifications
Specifications
Linearity Control Kit
5 Days Open, 120 Days Closed Vial
CELL DYN 3200 and hematology analyzer
CVA for WBC
5
WBC, platelet and RBC or Hemoglobin vials
5 x 3mL