Learn More
Streck CVA™ Calibration Verification Assessment/Linearity Kit
SDP
Ensuring confidence in every result, CVA (Calibration Verification Assessment) tests the extreme upper and lower limits of an instrument to assure abnormal patient results are valid
Supplier:  Streck 291720
 View more versions of this product
Catalog No. 11-716-368
Item Discontinued This item has been discontinued by the supplier. Please to view product availability in your area.
This item is not returnable. View return policy
Description
Description
Assayed linearity control kits to measure the linear performance of of three- and five-part hematology analyzers
  • Instruments include:
  • Beckman Coulter™, Ac-T™ 5diff, HmX, UniCel™ DxH TM800, LH 500, LH 750/755, LH 780/785; HemoCue™, Inc. Hb 201+ Analyzer; Mindray™ BC-3200; Abbott CELL-DYN™ Sapphire™, Ruby™, Emerald™, 3500/3700, 3200, 1800
  • Include customized ranges appropriate for each instrument model and provide instrument-specific assay values for the WBC, platelet, RBC and hemoglobin parameters
  • Open-vial stability is 5 days
  • Closed-vial stability is 120 days
  • All kits include WBC, platelet and RBC/hemoglobin vials unless otherwise noted
Specifications
Specifications
CELL DYN 3200 and hematology analyzer
WBC, platelet and RBC or Hemoglobin vials
5
5 Days Open, 120 Days Closed Vial
Linearity Control Kit
CVA for Platelets
5 x 3 mL