Streck CVA™ Calibration Verification Assessment/Linearity Kit

Ensuring confidence in every result, CVA (Calibration Verification Assessment) tests the extreme upper and lower limits of an instrument to assure abnormal patient results are valid

Manufacturer: Streck 291714

 View more versions of this product

Catalog No. 11-716-362

  Check Availability
Add to Cart

    Specifications

    Product Type Linearity control kit
    Stability 5 Days Open, 120 Days Closed Vial
    Includes WBC, platelet and RBC or Hemoglobin vials
    For Use With (Application) CVA for Platelets
    For Use With (Equipment) ADVIA™ 120 and LH750 hematology analyzer
    Quantity 6 x 3mL
    Level 5

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit