Learn More
Streck ESR-Auto Plus Analyzer
Automated 10-position ESR analyzer
Catalog Number | Mfr. No. | Product Type | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Catalog Number | Mfr. No. | Product Type | Price | Quantity & Availability | |||||
23-285-622
|
Streck
240316 |
ESR 657 mixer |
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. |
N/A |
23-285-620
|
Streck
240357 |
ESR-auto plus analyzer |
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. |
View substitute products » |
Description
- Accurately and precisely measures the sedimentation rate of erythrocytes in 1.2mL ESR-Vacuum Tubes
- Results measured in millimeters per hour (Westergren method) and available in 30 minutes
- Features random-access, built-in printer, sample ID capability and 15-minute prediction mode
- Incorporates a quality control system for monitoring the laboratory's quality control program; data can be downloaded to a Laboratory Information System
Specifications
LIS |