Learn More
Streck ESR-Auto Plus™ Analyzer Available on GSA/VA Contract for Federal Government customers only.
Automated 10-position ESR analyzer
Manufacturer:  Streck 240357
 View more versions of this product
Catalog No. 23-285-620
  Check Availability
Add to cart
Specifications Description & Specifications
Specifications
15 min.
1.2mL
ESR-auto plus analyzer
LIS
Designed to accurately and precisely measure the sedimentation rate of erythrocytes in ESR-vacuum tubes
110V
Accessories