Streck ESR-Chex™ Controls Available on GSA/VA Contract for Federal Government customers only.

Verify the precision and accuracy of both manual and automated ESR methods

Manufacturer: Streck 214024

 View more versions of this product

Catalog No. 23-046577

  Check Availability
Add to Cart

    Specifications

    Product Type ESR-Chex control
    Stability 95 Days Open, 12 Months Closed Vial
    Compatible Methods Both automated and manual ESR methods
    For Use With (Application) Verify the precision and accuracy of both manual and automated ESR methods
    For Use With (Equipment) Clinical chemistry analyzer
    Certifications/Compliance Internal quality control standards and regulatory compliance
    Shelf Life 1 Year
    Storage Requirements 2° to 10°C
    Quantity 4 x 9mL,4 x 9mL
    Level 2

  • Description & Specifications
  • Accessories

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit