Learn More
Streck ESR-Vacuum Tubes
For use with the Streck ESR-Auto Plus™ and ESR-100 Sedimentation Rate Analyzers
Manufacturer:  Streck 240360
 View more versions of this product
Catalog No. 11-716-338
Add to cart
Description
Description
Collect, transport, and analyze the Erythrocyte Sedimentation Rate (ESR) of whole blood in the clinical laboratory with Streck ESR-Vacuum Tubes. They are specifically designed for use with the Streck ESR-Auto Plus and ESR-100 automated sedimentation rate analyzers and the Streck ESR-10 Manual Rack.
  • For the Collection and Analysis of Whole Blood
  • Extended Stability: Patient samples can be maintained from the time of blood collection for up to 72 hours prior to analysis when transported and stored at 2-10°C in all Streck ESR-Vacuum Tubes
  • Tubes contain tri-sodium citrate to avoid coagulation
  • After the tube is mixed, it is placed in the ESR instrument; the instrument automatically prints or downloads the ESR result in 30 minutes
Safety Coated Tubes
  • Significantly more impact resistant than glass collection tubes
  • Designed with an outer Mylar™ coating that contains glass and blood specimens in the event of breakage
  • Reduces the risk of injury and potential exposure to blood borne pathogens
Specifications
Specifications
100 Box
30 min.
ESR Vacuum Tube
Tri Sodium citrate to avoid coagulation
2 to 10°C
1.2mL
Streck ESR-Auto Plus™ and ESR-100 Sedimentation rate analyzers