Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
Streck CD-Chex TdT™ Plus Assay

Click to view available options
Quantity:
1 x 1 mL
2 x 1 mL
Stability:
30 Days Open
90 Days Open
Description
CD-Chex TdT Plus is a positive procedural control that provides reference values for TdT, CD1a, CD2, CD4, CD5, CD7, CD8, CD34 and cytoplasmic CD3. This control contains several markers designated as rare flow antigens by CAP.
- Only flow cytometry control assayed for TdT, CD1a, CD34 and cytoplasmic CD3.
- Offers an abnormally high level of CD34: over 60% positive for CD34+.
- Compatible with BD Biosciences and Beckman Coulter flow cytometry systems.
- 30-day open-vial stability, 90-day closed-vial stability.
- Manufacturer established assay ranges – no need to verify range per CAP requirements.
- Access to STATS™, Streck’s free interlaboratory quality control program for peer group data comparison.
Specifications
Specifications
For Use With (Equipment) | BD Biosciences and Beckman Coulter flow cytometry systems |
Type | CD-Chex TdT™ Plus Assay |
Quantity | 1 x 1 mL |
Stability | 90 Days Open |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction