Streck Para 12™ Hematology Controls

Multi-parameter assayed hematology control for instruments offering a three-part differential

Manufacturer: Streck 218618

 View all options for this product

Catalog No. 11-716-39


This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.  View substitute products »


    Product Type Para 12™ hematology control
    Source Stabilized human or mammalian RBC, human, mammalian or simulated white blood cells and a platelet component
    Level 6 Low, 6 Normal, 6 High
    Format Vial
    Stability 110 Days
    For Use With (Application) For evaluating the accuracy and precision of hematology instruments that provide a WBC differential
    For Use With (Equipment) Beckman Coulter™ AC.T™ Series, AC T diff™, AC T diff2™, Abbott CELL-DYN™ 1400, 1600, 1700, and 1800, Horiba medical ABX micros 60, Mindray BC-3200, Siemens ADVIA™
    Certifications/Compliance CE
    Storage Requirements 2° to 10°C
    Quantity 18 x 4.5mL

    • Three distinct populations of lymphocytes, mononuclears, and granulocytes
    • Varying percentages offered in the low abnormal, normal, and high abnormal levels to test the accuracy and precision of the reported white cell populations
    • Values are recovered for all parameters ensuring all requirements are met and exceeded for complete laboratory quality control

  • Description & Specifications

Related Products

Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit