Streck Para 12™ Multiparameter Control

Multiparameter hematology control for instruments offering a three-part or five-part interpretive differential

Manufacturer: Streck 218503

 View all options for this product

Catalog No. 11-716-341


This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000.  View substitute products »


    Product Type Para 12™ multi parameter control
    Level Low, Normal and High
    Format Vial
    Stability 110 Days
    For Use With (Application) To test the accuracy and precision of white cell populations
    For Use With (Equipment) Beckman Coulter™ AC.T™ Series, AC T diff™, AC T diff2™, Abbott CELL-DYN™ 1400, 1600, 1700, and 1800, Horiba medical ABX micros 60, Mindray BC-3200, Siemens ADVIA™
    Certifications/Compliance CE
    Storage Requirements 2° to 10°C
    Quantity 2.5mL

    • Has three distinct populations of lymphocytes, mononuclears, and granulocytes
    • Varying percentages are also offered in the low-abnormal, normal and high-abnormal levels to test the accuracy and precision of the reported white cell populations
    • Values can be recovered for all parameters ensuring all requirements are met for complete laboratory quality control
    • Streck offers a free quality control program to all qualifying customers STATS™ provides convenient data submission options, quick turnaround time and relevant reports
    • Assayed for the following: Abbott CELL-DYN™ 1400, 1600, 1700 and 1800; ABX Micros 60; Beckman Coulter™, AcT and AcT diff Series

  • Description & Specifications

Related Products

Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit