Streck Para 12™ Plus Retics Control

Whole blood, five-part differential hematology control with reticulocytes added

Manufacturer: Streck 218954

 View more versions of this product

Catalog No. 11-716-326

 N/A
N/A

This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 


    Specifications

    Product Type Para 12™ plus retics control
    Format Vial
    Stability 75 Days
    For Use With (Application) For evaluating the accuracy and precision of hematology instruments that provide a WBC differential
    For Use With (Equipment) Abbott CELL-DYN™ five-part differential instrument
    Certifications/Compliance CE
    Storage Requirements 2° to 10°C
    Quantity 6 x 3.5mL,6 x 3.5mL
    Source Stabilized human or mammalian RBC, human, mammalian or simulated white blood cells and a platelet component,Stabilized human or mammalian RBC, human, mammalian or simulated white blood cells and a platelet component
    Level 2 Low, 2 Normal, 2 High,2 Low, 2 Normal, 2 High

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit