Streck Para 4™ Four Parameter Control

Designed for small laboratories performing a limited number of hematology tests, utilizing manual or semi-automated methods

Manufacturer: Streck 215408

 View more versions of this product

Catalog No. 11-716-281

  Check Availability
Add to Cart

    Specifications

    Product Type Para 4™ four parameter hematology control
    Format Glass Vial
    Stability 110 Days
    For Use With (Application) For small laboratories performing a limited number of hematology tests
    For Use With (Equipment) HemoCue™ B-Hemoglobin photometer and Hb201+, ITC Hgb Pro professional hemoglobin testing system
    Certifications/Compliance CE
    Storage Requirements 2° to 10°C
    Quantity 8 x 1.5mL,8 x 1.5mL
    Level 4 Low, 4 Normal,4 Low, 4 Normal
    Assay Target WBC, RBC, Hgb, Hct and Platelet

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit