Streck Para 4 Hematology Controls

Available in plastic dropper vials for easy sample allocation with hemoglobin and hematocrit instruments

Manufacturer: Streck 215106

 View more versions of this product

Catalog No. 23-285-631

  Check Availability
Add to Cart

    Specifications

    Product Type Para 4™ hematology control
    Format Plastic Vial
    Stability 110 Days
    For Use With (Application) For evaluating the accuracy and precision of automated, semi automated and manual procedures that measure Blood cell parameters
    For Use With (Equipment) HemoCue™ B-Hemoglobin photometer and Hb 201+ analyzer, HemataSTAT II™
    Certifications/Compliance CE
    Storage Requirements 2° to 10°C
    Quantity 6 x 1.5mL,6 x 1.5mL
    Source Stabilized human RBC, a white blood cell component consisting of human and/or non-human analogs and a platelet component,Stabilized human RBC, a white blood cell component consisting of human and/or non-human analogs and a platelet component
    Level 6 Low,6 Low

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit