Streck Para™ 5X Whole Blood Control

A whole blood hematology control manufactured specifically for the Horiba Medical ABX Pentra 60C+ and 80 and the Coulter™ Ac·T™ 5diff instruments

Manufacturer: Streck 215409

 View more versions of this product

Catalog No. 23-285-564

  Check Availability
Add to Cart

    Specifications

    Product Type Para™ 5X whole blood control
    Format Glass Vial
    Stability 110 Days
    Includes Assay disc
    For Use With (Application) White cell differential, positioning the populations within the limitations of the algorithms on the system
    For Use With (Equipment) Horiba medical ABX Pentra™ 60C+ and 80 and the Coulter™ Ac
    Quantity 8 x 1.5mL,8 x 1.5mL
    Level 4 Low, 4 High,4 Low, 4 High

  • Description & Specifications

Related Products


Product Content Feedback

We continue to work to improve your shopping experience and your feedback regarding this content is very important to us. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Cancel Submit