Streck Reagents for VC-100 Acid-Base Titration Pipet Verification Instrument
Acid-base titration method that verifies the volume delivered by pipets and automatic dilutors
Manufacturer: Streck 270250
 View more versions of this product
Catalog No. 11-716-215
 N/A
N/A
This item has been discontinued by the manufacturer and is no longer available. Please call customer service for assistance: 1-800-766-7000. 
    Specifications
    Includes 4 Bottles titration solution, 4 titration cartridges, 6 vials each A and B reference control, 14 vials titration acid, 150 titration cups, pipet labels
    For Use With VC-100 Acid base titration pipet verification instrument
    Applications Verifying the volume delivered by pipets and dilutors
    Type Pipet verification kit
  • Description & Specifications
Related Products
Product Content Feedback